accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13721-20-5

IUPAC Name: 2-(3-chloro-4-methoxyphenyl)acetic acid
Synonyms: (3-chloro-4-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: OARWXDXELDVNOC-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(CC(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results