accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH2NCl

CAS RN 13726-14-2

IUPAC Name: 4-chloro-3-methoxyaniline
Synonyms: 4-chloro-3-methoxyaniline
Molecular Weight (g/mol): 157.6
Molecular Formula: C7H8ClNO
InChi Key: LNKBDFVSILQKSI-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(N)=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)