accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13726-76-6

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-[(diaminomethylidene)amino]pentanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Weight (g/mol): 274.32
Molecular Formula: C11H22N4O4
InChi Key: HSQIYOPBCOPMSS-ZETCQYMHSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CCCN=C(N)N)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results