accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OO(S)HNOOCH3H3CCH3OO

CAS RN 13726-84-6

IUPAC Name: (2S)-5-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoate
Synonyms: (2S)-5-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]-5-oxopentanoate
Molecular Weight (g/mol): 302.35
Molecular Formula: C14H24NO6
InChi Key: YGSRAYJBEREVRB-VIFPVBQESA-M
SMILES: CC(C)(C)OC(=O)CC[C@H](NC(=O)OC(C)(C)C)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results