accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH3C

CAS RN 13728-34-2

IUPAC Name: 2,3-dimethyl naphthalene-2,3-dicarboxylate
Synonyms: 2,3-dimethyl naphthalene-2,3-dicarboxylate
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: MPDGBCOIHNLQMR-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=C2C=CC=CC2=C1)C(=O)OC
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)