accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 137281-39-1

IUPAC Name: 4-(2-{2-amino-4-oxo-1H,4H,7H-pyrrolo[2,3-d]pyrimidin-5-yl}ethyl)benzoic acid
Synonyms: 4-(2-{2-amino-4-oxo-1H,7H-pyrrolo[2,3-d]pyrimidin-5-yl}ethyl)benzoic acid
Molecular Weight (g/mol): 298.3
Molecular Formula: C15H14N4O3
InChi Key: AIZPFZIKHIJCQX-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C2=C(NC=C2CCC2=CC=C(C=C2)C(O)=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results