accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)OHO(S)HNH3COOOCH3CH3CH3H3CCH3CH3

CAS RN 13734-40-2

IUPAC Name: (2S,3R)-3-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms: (2S,3R)-3-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]butanoic acid|BOC-THR(TBU)-OH
Molecular Weight (g/mol): 275.35
Molecular Formula: C13H25NO5
InChi Key: LKRXXARJBFBMCE-BDAKNGLRSA-N
SMILES: C[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)