accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13735-13-2

IUPAC Name: 6-bromo-3,4-dihydro-2H-1-benzothiopyran-4-one
Synonyms: 6-bromo-2,3-dihydro-1-benzothiopyran-4-one
Molecular Weight (g/mol): 243.12
Molecular Formula: C9H7BrOS
InChi Key: SBNPYDJTVGRDAA-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(SCCC2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results