accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3N

CAS RN 13737-04-7

IUPAC Name: 2-(trimethylsilyl)pyridine
Synonyms: 2-(trimethylsilyl)pyridine
Molecular Weight (g/mol): 151.28
Molecular Formula: C8H13NSi
InChi Key: IBLKFJNZUSUTPB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results