CAS RN 13737-05-8
IUPAC Name:
2-(trimethylstannyl)pyridine
Synonyms:
2-(trimethylstannyl)pyridine
Molecular Weight (g/mol):
241.91
Molecular Formula:
C8H13NSn
InChi Key:
HDWHGDDNMJOCCU-UHFFFAOYSA-N
SMILES:
C[Sn](C)(C)C1=CC=CC=N1
1
–
1
of
1
results
1
Trimethyl(2-pyridyl)tin 98.0+%, TCI America™
CAS: 13737-05-8 Molecular Formula: C8H13NSn Molecular Weight (g/mol): 241.91 MDL Number: MFCD01631312 InChI Key: HDWHGDDNMJOCCU-UHFFFAOYSA-N Synonym: 2-Trimethylstannylpyridine PubChem CID: 11021057 IUPAC Name: 2-(trimethylstannyl)pyridine SMILES: C[Sn](C)(C)C1=CC=CC=N1