accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 137386-78-8

IUPAC Name: 3-(pyridin-2-yloxy)benzaldehyde
Synonyms: 3-(pyridin-2-yloxy)benzaldehyde
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: YGXYNKLOBXMTGL-UHFFFAOYSA-N
SMILES: O=CC1=CC(OC2=CC=CC=N2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results