accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1373920-66-1

IUPAC Name: 2-fluoro-5-methyl-4-(trifluoromethyl)aniline
Synonyms: 2-fluoro-5-methyl-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 193.15
Molecular Formula: C8H7F4N
InChi Key: SCCCZPNCTRGYDF-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results