accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFNH2F

CAS RN 1373920-83-2

IUPAC Name: 4-butoxy-2,6-difluoroaniline
Synonyms: 4-butoxy-2,6-difluoroaniline
Molecular Weight (g/mol): 201.22
Molecular Formula: C10H13F2NO
InChi Key: QJBXYVXNGAAECC-UHFFFAOYSA-N
SMILES: CCCCOC1=CC(F)=C(N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results