CAS RN 1373920-89-8
IUPAC Name:
4-chloro-3-ethoxy-2-fluorobenzonitrile
Synonyms:
4-chloro-3-ethoxy-2-fluorobenzonitrile
Molecular Weight (g/mol):
199.61
Molecular Formula:
C9H7ClFNO
InChi Key:
SVYHZDFUKRNQNH-UHFFFAOYSA-N
SMILES:
CCOC1=C(Cl)C=CC(C#N)=C1F
1
–
1
of
1
results
1
4-Chloro-3-ethoxy-2-fluorobenzonitrile, 97%, Thermo Scientific™
CAS: 1373920-89-8 Molecular Formula: C9H7ClFNO Molecular Weight (g/mol): 199.609 MDL Number: MFCD22201019 InChI Key: SVYHZDFUKRNQNH-UHFFFAOYSA-N PubChem CID: 86277623 IUPAC Name: 4-chloro-3-ethoxy-2-fluorobenzonitrile SMILES: CCOC1=C(C=CC(=C1F)C#N)Cl