accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13754-38-6

IUPAC Name: 4-benzoylpiperazin-1-ium
Synonyms: 4-benzoylpiperazin-1-ium
Molecular Weight (g/mol): 191.25
Molecular Formula: C11H15N2O
InChi Key: VUNXBQRNMNVUMV-UHFFFAOYSA-O
SMILES: O=C(N1CC[NH2+]CC1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)