accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaH2OH2OCNFeCNNCNCNNOC

CAS RN 13755-38-9

IUPAC Name: disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Synonyms: disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Molecular Weight (g/mol): 297.95
Molecular Formula: C5H4FeN6Na2O3
InChi Key: JFPDVRGGFBUORC-UHFFFAOYSA-N
SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Quantity 
  • (2)
  • (9)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (8)
  • (2)
  • (2)
  • (1)
  • (11)
  • (2)
Physical Form 
  • (4)
  • (1)
  • (12)
  • (4)
  • (1)
Grade 
  • (4)
  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (12)
  • (1)
  • (2)
  • (9)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (6)
  • (2)
  • (3)
  • (8)
  • (2)
  • (2)
  • (1)
  • (11)
  • (2)
  • (4)
  • (1)
  • (12)
  • (4)
  • (1)
  • (4)
  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)