accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOBr

CAS RN 137577-00-5

IUPAC Name: 2-bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)ethan-1-one
Synonyms: 2-bromo-1-(5-methyl-1-phenylpyrazol-4-yl)ethanone
Molecular Weight (g/mol): 279.14
Molecular Formula: C12H11BrN2O
InChi Key: VYGXRQSIPNGJNK-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1C1=CC=CC=C1)C(=O)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results