accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NCH3H3COCl

CAS RN 1375930-97-4

IUPAC Name: 4-(chloromethyl)-N,N-bis(propan-2-yl)benzamide
Synonyms: 4-(chloromethyl)-N,N-diisopropylbenzamide
Molecular Weight (g/mol): 253.77
Molecular Formula: C14H20ClNO
InChi Key: IWHDTZURUKJHFU-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)C1=CC=C(CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results