accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFTm

CAS RN 13760-79-7

IUPAC Name: thulium(3+) trifluoride
Synonyms: thulium(3+) trifluoride
Molecular Weight (g/mol): 225.93
Molecular Formula: F3Tm
InChi Key: AATUHDXSJTXIHB-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Tm+3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results