accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFLu

CAS RN 13760-81-1

IUPAC Name: lutetium(3+) trifluoride
Synonyms: lutetium(3+) trifluoride
Molecular Weight (g/mol): 231.96
Molecular Formula: F3Lu
InChi Key: VIHLFTMKXFWYAS-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Lu+3]
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)