accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaOCrOOO

CAS RN 13765-19-0

IUPAC Name: calcium dioxochromiumbis(olate)
Synonyms: calcium chromate
Molecular Weight (g/mol): 156.07
Molecular Formula: CaCrO4
InChi Key: RFAFBXGYHBOUMV-UHFFFAOYSA-N
SMILES: [Ca++].[O-][Cr]([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results