accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13765-74-7

IUPAC Name: disilver(1+) dioxomolybdenumbis(olate)
Synonyms: silver molybdate
Molecular Weight (g/mol): 375.68
Molecular Formula: Ag2MoO4
InChi Key: MHLYOTJKDAAHGI-UHFFFAOYSA-N
SMILES: [Ag+].[Ag+].[O-][Mo]([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results