accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 137682-89-4

IUPAC Name: ethyl 1-hydroxycyclopropane-1-carboxylate
Synonyms: ethyl 1-hydroxycyclopropane-1-carboxylate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: DPMGQZGXWRHYPE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(O)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results