accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgNH2SOOONH2SOOO

CAS RN 13770-91-7

IUPAC Name: magnesium(2+) disulfamate
Synonyms: magnesium(2+) disulfamate
Molecular Weight (g/mol): 216.47
Molecular Formula: H4MgN2O6S2
InChi Key: YZVJHCGMTYDKFR-UHFFFAOYSA-L
SMILES: [Mg++].NS([O-])(=O)=O.NS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)