accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13777-25-8

IUPAC Name: zirconium(4+) tetrabromide
Synonyms: zirconium(IV) bromide
Molecular Weight (g/mol): 410.84
Molecular Formula: Br4Zr
InChi Key: LSWWNKUULMMMIL-UHFFFAOYSA-J
SMILES: [Br-].[Br-].[Br-].[Br-].[Zr+4]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results