accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13780-48-8

IUPAC Name: λ²-europium(2+) dibromide
Synonyms: λ²-europium(2+) dibromide
Molecular Weight (g/mol): 311.77
Molecular Formula: Br2Eu
InChi Key: PJVPGMOCWCUQHP-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Eu++]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results