accessibility menu, dialog, popup

UserName

Viewing profile as user:
SI

CAS RN 13781-37-8

IUPAC Name: 2-iodo-5-phenylthiophene
Synonyms: 2-iodo-5-phenylthiophene
Molecular Weight (g/mol): 286.13
Molecular Formula: C10H7IS
InChi Key: DHRIQSGKMRCNIU-UHFFFAOYSA-N
SMILES: IC1=CC=C(S1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results