accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SFFFFFFSH3C

CAS RN 137814-07-4

IUPAC Name: 3-[3,3,4,4,5,5-hexafluoro-2-(2-methyl-1-benzothiophen-3-yl)cyclopent-1-en-1-yl]-2-methyl-1-benzothiophene
Synonyms: 3-[3,3,4,4,5,5-hexafluoro-2-(2-methyl-1-benzothiophen-3-yl)cyclopent-1-en-1-yl]-2-methyl-1-benzothiophene
Molecular Weight (g/mol): 468.48
Molecular Formula: C23H14F6S2
InChi Key: CNLMHUFAXSWHFA-UHFFFAOYSA-N
SMILES: CC1=C(C2=CC=CC=C2S1)C1=C(C2=C(C)SC3=CC=CC=C23)C(F)(F)C(F)(F)C1(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)