accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 137829-79-9

IUPAC Name: 5,11-di-tert-butyl-2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Synonyms: 5,11-di-tert-butyl-2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Molecular Weight (g/mol): 374.33
Molecular Formula: C22H33BF2N2
InChi Key: SEHGNHOGQDPQRC-UHFFFAOYSA-N
SMILES: CC1=C(C(C)=C2N1[B-](F)(F)[N+]1=C(C)C(=C(C)C1=C2C)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)