accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 137911-28-5

IUPAC Name: 4-({4'-[(4-formylphenyl)(phenyl)amino]-[1,1'-biphenyl]-4-yl}(phenyl)amino)benzaldehyde
Synonyms: 4-({4'-[(4-formylphenyl)(phenyl)amino]-[1,1'-biphenyl]-4-yl}(phenyl)amino)benzaldehyde
Molecular Weight (g/mol): 544.65
Molecular Formula: C38H28N2O2
InChi Key: DMVJDNDXIBORDT-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results