accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13794-72-4

IUPAC Name: 6,7-dimethoxy-1,4-dihydroquinazolin-4-one
Synonyms: 6,7-dimethoxy-1H-quinazolin-4-one
Molecular Weight (g/mol): 206.2
Molecular Formula: C10H10N2O3
InChi Key: DMSRMHGCZUXCMJ-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(=O)N=CNC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)