CAS RN 13795-24-9
CAS RN 13795-24-9
IUPAC Name:
benzyl 4-nitrophenyl carbonate
Synonyms:
benzyl 4-nitrophenyl carbonate
Molecular Weight (g/mol):
273.24
Molecular Formula:
C14H11NO5
InChi Key:
QIXRWIVDBZJDGD-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(OC(=O)OCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
Benzyl 4-Nitrophenyl Carbonate 96.0+%, TCI America™
CAS: 13795-24-9 Molecular Formula: C14H11NO5 Molecular Weight (g/mol): 273.24 MDL Number: MFCD00007323 InChI Key: QIXRWIVDBZJDGD-UHFFFAOYSA-N Synonym: Carbonic Acid Benzyl 4-Nitrophenyl Ester PubChem CID: 83737 IUPAC Name: benzyl 4-nitrophenyl carbonate SMILES: [O-][N+](=O)C1=CC=C(OC(=O)OCC2=CC=CC=C2)C=C1