accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13799-90-1

IUPAC Name: 2,5-dichlorobenzene-1,4-dicarboxylic acid
Synonyms: 2,5-dichloro-terephthalic acid
Molecular Weight (g/mol): 235.02
Molecular Formula: C8H4Cl2O4
InChi Key: LMOSYFZLPBHEOW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=C(C=C1Cl)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results