accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138-15-8

IUPAC Name: hydrogen 2-aminopentanedioic acid chloride
Synonyms: hydrogen glutaminsaeure chloride
Molecular Weight (g/mol): 183.59
Molecular Formula: C5H10ClNO4
InChi Key: RPAJSBKBKSSMLJ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC(CCC(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)