accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138-22-7

CAS RN 138-22-7

IUPAC Name: butyl 2-hydroxypropanoate
Synonyms: butyl lactate N-butyl lactate
Molecular Weight (g/mol): 146.19
Molecular Formula: C7H14O3
InChi Key: MRABAEUHTLLEML-UHFFFAOYNA-N
SMILES: CCCCOC(=O)C(C)O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)