accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138-24-9

IUPAC Name: N,N,N-trimethylanilinium chloride
Synonyms: tetrabutylammonium tribromide chloride
Molecular Weight (g/mol): 171.67
Molecular Formula: C9H14ClN
InChi Key: MQAYPFVXSPHGJM-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)