CAS RN 138-85-2
CAS RN 138-85-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
361.7
Molecular Formula:
C7H6HgNaO3
InChi Key:
BERBQUNVJGVZCX-UHFFFAOYSA-M
SMILES:
O.[Na+].[O-]C(=O)C1=CC=C([Hg])C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 4-(Hydroxymercury)benzoic Acid Sodium Salt, 98%
CAS: 138-85-2 Molecular Formula: C7H5HgNaO3 Molecular Weight (g/mol): 360.69 MDL Number: MFCD00002527 InChI Key: BERBQUNVJGVZCX-UHFFFAOYSA-M Synonym: sodium 4-carboxylatophenyl mercury hydrate,sodium hydrate p-mercuribenzoate PubChem CID: 23667696 IUPAC Name: sodium;(4-carboxylatophenyl)mercury;hydrate SMILES: C1=CC(=CC=C1C(=O)[O-])[Hg].O.[Na+]