accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138-86-3

IUPAC Name: 1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Synonyms: limonene, (+)- 1,8-p-menthadiene D,L-limonene 1,8(9)-p-menthadiene S-(-)-limonene polydipentene dl-limonene (racemic) p-mentha-1,8-diene (dipentene) terpenes and terpenoids
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: XMGQYMWWDOXHJM-UHFFFAOYNA-N
SMILES: CC(=C)C1CCC(C)=CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results