accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13803-74-2

IUPAC Name: 4-methylhexan-2-amine hydrochloride
Synonyms: methylhexanamine hydrochloride
Molecular Weight (g/mol): 151.68
Molecular Formula: C7H18ClN
InChi Key: ZKKBPHUAHARETG-UHFFFAOYNA-N
SMILES: Cl.CCC(C)CC(C)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results