accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONN

CAS RN 1380500-94-6

IUPAC Name: tert-butyl 2-cyano-1H-pyrrole-1-carboxylate
Synonyms: tert-butyl 2-cyanopyrrole-1-carboxylate
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: GDHNTBXGTBFNIC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results