accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1380571-72-1

IUPAC Name: bis(6-oxa-1-azaspiro[3.3]heptane); oxalic acid
Synonyms: bis(6-oxa-1-azaspiro[3.3]heptane); oxalic acid
Molecular Weight (g/mol): 288.3
Molecular Formula: C12H20N2O6
InChi Key: UMKDEMSXCWMHRS-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.C1CC2(COC2)N1.C1CC2(COC2)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results