accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNH2HClHCl

CAS RN 138060-07-8

IUPAC Name: piperidin-3-amine dihydrochloride
Synonyms: piperidin-3-amine dihydrochloride
Molecular Weight (g/mol): 173.08
Molecular Formula: C5H14Cl2N2
InChi Key: GGPNYXIOFZLNKW-UHFFFAOYNA-N
SMILES: Cl.Cl.NC1CCCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results