CAS RN 138109-49-6
CAS RN 138109-49-6
IUPAC Name:
magnesium(2+) 2,4-dimethoxybenzen-1-ide bromide
Synonyms:
magnesium(2+) 2,4-dimethoxybenzen-1-ide bromide
Molecular Weight (g/mol):
241.37
Molecular Formula:
C8H9BrMgO2
InChi Key:
LWSQHQVQBSQPJQ-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].COC1=CC=[C-]C(OC)=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-Hydroxy-L-Phenylglycine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.