CAS RN 13813-40-6
IUPAC Name:
terbium(3+) triiodide
Synonyms:
terbium(III) iodide
Molecular Weight (g/mol):
539.64
Molecular Formula:
I3Tb
InChi Key:
OJXRJPFRTRETRN-UHFFFAOYSA-K
SMILES:
[I-].[I-].[I-].[Tb+3]
1
–
1
of
1
results
1
Terbium(III) iodide, ultra dry, 99.99% (REO)
CAS: 13813-40-6 MDL Number: MFCD00049573