accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13815-39-9

IUPAC Name: platinum(2+) dipotassium tetranitrite
Synonyms: platinum(2+) dipotassium tetranitrite
Molecular Weight (g/mol): 457.3
Molecular Formula: K2N4O8Pt
InChi Key: GQUAVGOGNMMUFS-UHFFFAOYSA-J
SMILES: [K+].[K+].[Pt++].[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results