accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138171-14-9

IUPAC Name: N1,N4-bis(4-methylphenyl)-N1,N4-diphenylbenzene-1,4-diamine
Synonyms: N1,N4-bis(4-methylphenyl)-N1,N4-diphenylbenzene-1,4-diamine
Molecular Weight (g/mol): 440.59
Molecular Formula: C32H28N2
InChi Key: FQNVFRPAQRVHKO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results