accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrBrGd

CAS RN 13818-75-2

IUPAC Name: gadolinium(3+) tribromide
Synonyms: gadolinium(3+) tribromide
Molecular Weight (g/mol): 396.96
Molecular Formula: Br3Gd
InChi Key: KGOKDPWKDBWITQ-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Gd+3]
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)