accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(S)OHNCl(R)OH(S)HO(R)HO

CAS RN 138182-21-5

IUPAC Name: (2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: (2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 329.73
Molecular Formula: C14H16ClNO6
InChi Key: OQWBAXBVBGNSPW-MBJXGIAVSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)