accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13819-84-6

IUPAC Name: molybdenum(5+) pentafluoride
Synonyms: molybdenum(V) fluoride
Molecular Weight (g/mol): 190.94
Molecular Formula: F5Mo
InChi Key: NBJFDNVXVFBQDX-UHFFFAOYSA-I
SMILES: [F-].[F-].[F-].[F-].[F-].[Mo+5]
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results