accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOH

CAS RN 138194-04-4

IUPAC Name: [5-(pyridin-4-yl)thiophen-2-yl]methanol
Synonyms: [5-(pyridin-4-yl)thiophen-2-yl]methanol
Molecular Weight (g/mol): 191.25
Molecular Formula: C10H9NOS
InChi Key: GDGUQNXUEMPEKM-UHFFFAOYSA-N
SMILES: OCC1=CC=C(S1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results